Skip to Content
Products
Cart
0
AS
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Testing
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Greiner CELLSTAR
®
dish
Synonym(s):
Greiner CELLSTAR
®
cell culture dishes
Products
Product Category
dishes (4)
Brand
Sigma-Aldrich (1)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Surface Area cm²
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P7612
Greiner CELLSTAR
®
dish,
diam. × H 100 mm × 20 mm, vented
Expand
View Pricing
P7737
Greiner CELLSTAR
®
dish,
diam. × H 145 mm × 20 mm, vented
Expand
View Pricing
P6987
Greiner CELLSTAR
®
dish,
diam. × H 35 mm × 10 mm, vented
Expand
View Pricing
P7237
Greiner CELLSTAR
®
dish,
diam. × H 60 mm × 15 mm, vented
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More