Skip to Content
MilliporeSigma
Gold(III) chloride trihydrate Technipur®, ≥99.9% trace metals basis

Gold(III) chloride trihydrate

Gold(III) chloride trihydrate Technipur®, ≥99.9% trace metals basis
Synonym(s):
Hydrogen tetrachloroaurate(III), Tetrachloroauric(III) acid
Linear Formula:
HAuCl4 · 3H2O
CAS Number:
Molecular Weight:
393.83

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Package Size
Reagent Type
Core
Special Grade