Skip to Content
MilliporeSigma
Glufosinate-ammonium certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Glufosinate-ammonium

Glufosinate-ammonium certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-Amino-4-(hydroxymethylphosphinyl)butyric acid ammonium salt, DL-Phosphinothricin
Empirical Formula (Hill Notation):
C5H15N2O4P
CAS Number:
Molecular Weight:
198.16
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique