Skip to Content
MilliporeSigma
Ginsenoside Rg1 primary reference standard

Ginsenoside Rg1

Ginsenoside Rg1 primary reference standard
Synonym(s):
(3β,6α,12β)-3,12-Dihydroxydammar-24-ene-6,20-diyl bis-β-D-glucopyranoside, Ginsenoside A2, Ginsenoside g1, Panaxoside A, Panaxoside Rg1, Sanchinoside C1, Sanchinoside Rg1, (3β,6α,12β)-3,12-Dihydroxydammar-24-ene-6,20-diyl bis-β-D-glucopyranoside, Panaxsaponin Rg1, Rg1 ginsenoside, Ginsenoside Rg1
Empirical Formula (Hill Notation):
C42H72O14
CAS Number:
Molecular Weight:
801.01
EC Number:

Products

Brand
Feature
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique