Skip to Content
MilliporeSigma
Ginsenoside Rd analytical standard

Ginsenoside Rd

Ginsenoside Rd analytical standard
Synonym(s):
(3β,12β)-20-(β-D-Glucopyranosyloxy)-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, Gypenoside VIII, (3β,12β)-20-(β-D-Glucopyranosyloxy)-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, 3-O-β-D-Glucopyranosyl-(1→2)-β-D-glucopyranosyl-20-O-β-D-glucopyranosyldammar-24-ene-3β,12β,20S-triol, Chikusetsusaponin FK7, Ginsenoside Rd1, Gypenoside VIII
Empirical Formula (Hill Notation):
C48H82O18
CAS Number:
Molecular Weight:
947.15

Products

Brand
Color
Feature
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing