Skip to Content
MilliporeSigma
Ginsenoside Rb2 analytical standard

Ginsenoside Rb2

Ginsenoside Rb2 analytical standard
Synonym(s):
(3β,12β)-20-[(6-O-α-L-Arabinopyranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, NSC 308878
Empirical Formula (Hill Notation):
C53H90O22
CAS Number:
Molecular Weight:
1079.27
EC Number:

Products

Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing