Skip to Content
MilliporeSigma
Furosemide British Pharmacopoeia (BP) Reference Standard

Furosemide

Furosemide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfonyl)-4-chloro-2-([2-furanylmethyl]amino)benzoic acid
Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique