Skip to Content
MilliporeSigma
Fmoc-His(Trt)-OH ≥98.0% (sum of enantiomers, HPLC)

Fmoc-His(Trt)-OH

Fmoc-His(Trt)-OH ≥98.0% (sum of enantiomers, HPLC)
Synonym(s):
Nα-Fmoc-N(im)-trityl-L-histidine, Fmoc-His(Trt)-OH
Empirical Formula (Hill Notation):
C40H33N3O4
CAS Number:
Molecular Weight:
619.71

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2