Skip to Content
MilliporeSigma
vibrant-m

Fluoxetine impurity B

vibrant-m
Synonym(s):
N-Methyl-N-(3-phenylpropyl)amine
Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing