Skip to Content
MilliporeSigma
Fluoroethylene carbonate Technipur®, 99% (GC)

Fluoroethylene carbonate

Fluoroethylene carbonate Technipur®, 99% (GC)
Synonym(s):
4-Fluoro-1,3-dioxolan-2-one, FEC
Empirical Formula (Hill Notation):
C3H3FO3
CAS Number:
Molecular Weight:
106.05

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade