Skip to Content
MilliporeSigma
Fluazuron PESTANAL®, analytical standard

Fluazuron

Fluazuron PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C20H10Cl2F5N3O3
CAS Number:
Molecular Weight:
506.21

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing