Skip to Content
MilliporeSigma
Ethyl malonyl chloride technical grade

Ethyl malonyl chloride

Ethyl malonyl chloride technical grade
Synonym(s):
Ethyl (chloroformyl)acetate, Ethyl 3-chloro-3-oxopropionate
Linear Formula:
CH3CH2OCOCH2COCl
CAS Number:
Molecular Weight:
150.56
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing