Skip to Content
MilliporeSigma
Ethyl isobutyrate natural, ≥98%, FCC, FG

Ethyl isobutyrate

Ethyl isobutyrate natural, ≥98%, FCC, FG
Linear Formula:
(CH3)2CHCOOC2H5
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing