Skip to Content
MilliporeSigma
Ethylhexyl triazone analytical standard

Ethylhexyl Triazone

Ethylhexyl triazone analytical standard
Synonym(s):
Ethylhexyl triazone, 2,4,6-Trianilino(p-carbo-2-ethylhexyl-1-oxy)-1,3,5-triazine, Tris(2-ethylhexyl) 4,4′,4″-(1,3,5-triazine-2,4,6-triyltriimino)trisbenzoate
Empirical Formula (Hill Notation):
C48H66N6O6
CAS Number:
Molecular Weight:
823.07
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing