Skip to Content
MilliporeSigma
Ethyldiphenylphosphine 98%

Ethyldiphenylphosphine

Ethyldiphenylphosphine 98%
Synonym(s):
Diphenylethylphosphine, NSC 151254
Linear Formula:
(C6H5)2PCH2CH3
CAS Number:
Molecular Weight:
214.24

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing