Skip to Content
MilliporeSigma
Ethyl acrylate SAJ first grade, ≥99.0%

Ethyl acrylate

Ethyl acrylate SAJ first grade, ≥99.0%
Synonym(s):
Acrylic acid ethyl ester, Ethyl acrylate
Linear Formula:
CH2=CHCOOC2H5
CAS Number:
Molecular Weight:
100.12
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Monomer Type
Reference Material Type
Special Grade
Technique