Skip to Content
MilliporeSigma
Ethidium bromide solution 10 mg/mL

Ethidium bromide solution

Ethidium bromide solution 10 mg/mL
Synonym(s):
3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, EtBr, Homidium bromide
Empirical Formula (Hill Notation):
C21H20BrN3
CAS Number:
Molecular Weight:
394.31

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Technique