
Dorzolamide impurity D

Synonym(s):
(4S,6S)-4-Amino-6-methyl-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide 7,7-dioxide hydrochloride, (4S,6S)-4-Amino-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide hydrochloride, N-Deethyldorzolamide hydrochloride
Empirical Formula (Hill Notation):
C8H12N2O4S3 · HCl
CAS Number:
Molecular Weight:
332.85
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing