
(±)-Nicotine

Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing