Skip to Content
MilliporeSigma
(±)-Miconazole nitrate salt Pharmaceutical Secondary Standard; Certified Reference Material

(±)-Miconazole nitrate salt

(±)-Miconazole nitrate salt Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
1-(2,4-Dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl)imidazole
Empirical Formula (Hill Notation):
C18H14Cl4N2O · HNO3
CAS Number:
Molecular Weight:
479.14
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique