Skip to Content
MilliporeSigma
(±)-DOI hydrochloride ≥98% (HPLC), solid

(±)-DOI hydrochloride

(±)-DOI hydrochloride ≥98% (HPLC), solid
Synonym(s):
(±)-1-(2,5-Dimethoxy-4-iodophenyl)-2-aminopropane hydrochloride, (±)-2,5-Dimethoxy-4-iodoamphetamine hydrochloride, (±)-DOI
Empirical Formula (Hill Notation):
C11H16INO2 · HCl
CAS Number:
Molecular Weight:
357.62

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing