Skip to Content
MilliporeSigma
DL-4-Hydroxy-2-ketoglutaric acid lithium salt ≥90.0% (TLC)

DL-4-Hydroxy-2-ketoglutaric acid lithium salt

DL-4-Hydroxy-2-ketoglutaric acid lithium salt ≥90.0% (TLC)
Synonym(s):
4-Hydroxy-2-oxoglutaric acid lithium salt, DL-KHG Li
Empirical Formula (Hill Notation):
C5H6O6 · xLi+
CAS Number:
Molecular Weight:
162.10 (free acid basis)

Products

Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing