Skip to Content
MilliporeSigma
Dithranol British Pharmacopoeia (BP) Reference Standard

Dithranol

Dithranol British Pharmacopoeia (BP) Reference Standard
Synonym(s):
1,8,9-Anthracenetriol, 1,8-Dihydroxy-9(10H)-anthracenone, 1,8-Dihydroxyanthrone, Anthralin
Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-3 of 3