Skip to Content
MilliporeSigma
DIPSO BioXtra, pH 4.0-5.5 (20 °C, 0.1 M in H2O), ≥98% (titration)

DIPSO

DIPSO BioXtra, pH 4.0-5.5 (20 °C, 0.1 M in H2O), ≥98% (titration)
Synonym(s):
3-(N,N-Bis[2-hydroxyethyl]amino)-2-hydroxypropanesulfonic acid, N,N-Bis(2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid
Empirical Formula (Hill Notation):
C7H17NO6S
CAS Number:
Molecular Weight:
243.28
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing