Skip to Content
MilliporeSigma
Di(propylene glycol) methyl ether acetate, mixture of isomers 99%

Di(propylene glycol) methyl ether acetate, mixture of isomers

Di(propylene glycol) methyl ether acetate, mixture of isomers 99%
Linear Formula:
CH3CO2C3H6OC3H6OCH3
CAS Number:
Molecular Weight:
190.24

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Polymer Composition
Polymer Type