Skip to Content
MilliporeSigma
Diphenyleneiodonium chloride ≥98%

Diphenyleneiodonium chloride

Diphenyleneiodonium chloride ≥98%
Synonym(s):
DPI, Dibenziodolium chloride, Diphenyleneiodonium Chloride
Empirical Formula (Hill Notation):
C12H8ClI
CAS Number:
Molecular Weight:
314.55

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment