Skip to Content
MilliporeSigma
Dimethyl 3,4-furandicarboxylate 98%

Dimethyl 3,4-furandicarboxylate

Dimethyl 3,4-furandicarboxylate 98%
Empirical Formula (Hill Notation):
C8H8O5
CAS Number:
Molecular Weight:
184.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing