Skip to Content
MilliporeSigma
Dihexylamine 97%

Dihexylamine

Dihexylamine 97%
Synonym(s):
Dihexylamine
Linear Formula:
CH3(CH2)5NH(CH2)5CH3
CAS Number:
Molecular Weight:
185.35

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing