Skip to Content
MilliporeSigma
DiFMUP ≥98% (HPLC)

DiFMUP

DiFMUP ≥98% (HPLC)
Synonym(s):
6,8-Difluoro-4-methylumbelliferyl phosphate
Empirical Formula (Hill Notation):
C10H7F2O6P
CAS Number:
Molecular Weight:
292.13

Products

Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing