Skip to Content
MilliporeSigma
Difluoromethyl 2-pyridyl sulfone 97% (HPLC)

Difluoromethyl 2-pyridyl sulfone

Difluoromethyl 2-pyridyl sulfone 97% (HPLC)
Synonym(s):
2-(Difluoromethylsulfonyl)pyridine, 2-PySO2CF2H
Empirical Formula (Hill Notation):
C6H5F2NO2S
CAS Number:
Molecular Weight:
193.17

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing