Skip to Content
AS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Testing
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diethylzinc
Synonym(s):
DEZ, Zincdiethyl
Linear Formula:
(C
2
H
5
)
2
Zn
CAS Number:
557-20-0
Molecular Weight:
123.51
EC Number:
209-161-3
Products
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
256781
Diethylzinc,
≥52 wt. % Zn basis
Expand
View Pricing
668729
Diethylzinc,
packaged for use in deposition systems
Expand
View Pricing