Skip to Content
MilliporeSigma
Diethylenetriamine SAJ first grade, ≥98.0%

Diethylenetriamine

Diethylenetriamine SAJ first grade, ≥98.0%
Synonym(s):
2,2′-Diaminodiethylamine, 2,2′-Iminodiethylamine, Bis(2-aminoethyl)amine, Diethylenetriamine
Linear Formula:
(NH2CH2CH2)2NH
CAS Number:
Molecular Weight:
103.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-4 of 4