Skip to Content
MilliporeSigma
Diethyl [difluoro(trimethylsilyl)methyl]phosphonate ≥85.0% (H-NMR)

Diethyl [difluoro(trimethylsilyl)methyl]phosphonate

Diethyl [difluoro(trimethylsilyl)methyl]phosphonate ≥85.0% (H-NMR)
Linear Formula:
(CH3)3SiCF2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
260.29

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing