Skip to Content
MilliporeSigma
Diethanolamine SAJ first grade, ≥98.0%

Diethanolamine

Diethanolamine SAJ first grade, ≥98.0%
Synonym(s):
2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine, Diethanolamine
Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique