Skip to Content
MilliporeSigma
Dicyclohexano-18-crown-6 98%

Dicyclohexano-18-crown-6

Dicyclohexano-18-crown-6 98%
Synonym(s):
2,3,11,12-Dicyclohexano-1,4,7,10,13,16-hexaoxacyclooctadecane, 2,5,8,15,18,21-Hexaoxatricyclo[20.4.0.09.14]hexacosane, Dicyclohexyl-18-crown-6, Perhydro-dibenzo-18-crown-6
Empirical Formula (Hill Notation):
C20H36O6
CAS Number:
Molecular Weight:
372.50
EC Number:

Products

Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing