Skip to Content
MilliporeSigma
Dichloromethane solution contains 10 % (v/v) methanol

Dichloromethane solution

Dichloromethane solution contains 10 % (v/v) methanol
Synonym(s):
Methylene chloride solution
Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing