Skip to Content
Merck
Dibenzylamine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Dibenzylamine

Dibenzylamine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Linear Formula:
(C6H5CH2)2NH
CAS Number:
Molecular Weight:
197.28

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing