
Dextrorphan tartrate

Synonym(s):
(+)-3-Hydroxy-N-methylmorphinan (+)-tartrate salt, Dextrorphan D-tartrate, Dextrorphan tartrate
Empirical Formula (Hill Notation):
C17H23NO · C4H6O6
CAS Number:
Molecular Weight:
407.46
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Application
Package Size
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing