Skip to Content
MilliporeSigma
D-(+)-Xylose 99%, natural sourced

D-(+)-Xylose

D-(+)-Xylose 99%, natural sourced
Synonym(s):
D(+)-Xylose
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Tissue
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Technique
Showing 1-6 of 6