Skip to Content
MilliporeSigma
D-(+)-Malic acid unnatural form, ≥97.0% (T)

D-(+)-Malic acid

D-(+)-Malic acid unnatural form, ≥97.0% (T)
Synonym(s):
D-(+)-Malic acid, (R)-(+)-2-Hydroxysuccinic acid, D-Hydroxybutanedioic acid
Linear Formula:
HOOCCH(OH)CH2COOH
CAS Number:
Molecular Weight:
134.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Package Size
API Family
Special Grade