Skip to Content
MilliporeSigma
D-(+)-Glucuronic acid γ-lactone analytical standard

D-(+)-Glucuronic acid γ-lactone

D-(+)-Glucuronic acid γ-lactone analytical standard
Synonym(s):
D-(+)-Glucurono-6,3-lactone, D-Glucurone, D-Glucurono-6,3-lactone, Glucuronolactone
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing