Skip to Content
MilliporeSigma
D-(+)-Glucose monohydrate tested according to Ph. Eur.

D-(+)-Glucose monohydrate

D-(+)-Glucose monohydrate tested according to Ph. Eur.
Synonym(s):
Dextrose monohydrate
Empirical Formula (Hill Notation):
C6H12O6 · H2O
CAS Number:
Molecular Weight:
198.17
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Optical Isomer