Skip to Content
MilliporeSigma
D-(+)-Galacturonic acid monohydrate analytical standard

D-(+)-Galacturonic acid monohydrate

D-(+)-Galacturonic acid monohydrate analytical standard
Empirical Formula (Hill Notation):
C6H10O7 · H2O
CAS Number:
Molecular Weight:
212.15
EC Number:

Products

Product Category
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2