Skip to Content
MilliporeSigma
D-Cysteine ≥99% (RT)

D-Cysteine

D-Cysteine ≥99% (RT)
Synonym(s):
(S)-2-Amino-3-mercaptopropionic acid
Empirical Formula (Hill Notation):
C3H7NO2S
CAS Number:
Molecular Weight:
121.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing