Skip to Content
MilliporeSigma
(+)-2,3-Dibenzoyl-D-tartaric acid ≥99.0% (T)

(+)-2,3-Dibenzoyl-D-tartaric acid

(+)-2,3-Dibenzoyl-D-tartaric acid ≥99.0% (T)
Synonym(s):
(+)-O,O′-Dibenzoyl-D-tartaric acid, D-Tartaric acid 2,3-dibenzoate
Linear Formula:
[C6H5CO2CH(CO2H)]2
CAS Number:
Molecular Weight:
358.30
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2