Skip to Content
MilliporeSigma
Cyclopentylphenylacetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Cyclopentylphenylacetic acid

Cyclopentylphenylacetic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Linear Formula:
C5H9CH(C6H5)CO2H
CAS Number:
Molecular Weight:
204.26

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade