Skip to Content
MilliporeSigma
Crystal Violet SAJ first grade

Crystal Violet

Crystal Violet SAJ first grade
Synonym(s):
Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B
Linear Formula:
C25H30N3Cl
CAS Number:
Molecular Weight:
407.98
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
Special Grade
Technique