Skip to Content
MilliporeSigma
Cresyl Violet acetate certified by the BSC

Cresyl Violet acetate

Cresyl Violet acetate certified by the BSC
Synonym(s):
9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt
Empirical Formula (Hill Notation):
C18H15N3O3
CAS Number:
Molecular Weight:
321.33
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Agency
Chemical Composition
Application
Technique
Showing 1-1 of 1