Skip to Content
MilliporeSigma
Corosolic acid from Lagerstroemia speciosa ≥98% (HPLC)

Corosolic acid from Lagerstroemia speciosa

Corosolic acid from Lagerstroemia speciosa ≥98% (HPLC)
Synonym(s):
Corosolic acid, (2α,3β)-2,3-Dihydroxyurs-12-en-28-oic acid, 2α-Hydroxyursolic acid, Colosolic acid, Corsolic acid, Glucosol
Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing