Skip to Content
MilliporeSigma
Copper(II) chloride dihydrate JIS special grade, ≥99.0%

Copper(II) chloride dihydrate

Copper(II) chloride dihydrate JIS special grade, ≥99.0%
Synonym(s):
Cupric chloride dihydrate, Copper(II) chloride dihydrate
Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reagent Type
Core
Special Grade
Technique